Skip to Content
Merck
All Photos(1)

Documents

SML0234

Sigma-Aldrich

Mitiglinide calcium

≥98% (HPLC)

Synonym(s):

(aS,3aR,7aS)-Octahydro-g-oxo-a-(phenylmethyl)-2H-isoindole-2-butanoic acid calcium salt, KAD 1229, Mitiglinide hemicalcium salt, S 21403, [2(S)-cis]-Octahydro-g-oxo-a-(phenylmethyl)-2H-isoindole-2-butanoic acid calclium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24NO3·0.5Ca
CAS Number:
Molecular Weight:
334.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +5 to +9° (c=1, MeOH)

storage condition

desiccated

color

white to beige

solubility

DMSO: ≥10 mg/mL at warmed to 60 °C

storage temp.

room temp

SMILES string

O=C(C[C@H](Cc1ccccc1)C(=O)O[Ca]OC(=O)[C@H](CC(=O)N2C[C@H]3CCCC[C@H]3C2)Cc4ccccc4)N5C[C@H]6CCCC[C@H]6C5

InChI

1S/2C19H25NO3.Ca/c2*21-18(20-12-15-8-4-5-9-16(15)13-20)11-17(19(22)23)10-14-6-2-1-3-7-14;/h2*1-3,6-7,15-17H,4-5,8-13H2,(H,22,23);/q;;+2/p-2/t2*15-,16+,17-;/m00./s1

InChI key

PMRVFZXOCRHXFE-FMEJWYFOSA-L

Biochem/physiol Actions

Mitiglinide is a drug for the treatment of type 2 diabetes. It is a highly selective KATP channel antagonist Mitiglinide is a hypoglycemic agent that closes adenosine triphosphate (ATP)–sensitive potassium channels (KATP channel s) in the pancreatic β-islet cells.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Jin et al.
Arzneimittel-Forschung, 62(1), 40-45 (2012-02-15)
A sensitive and specific method was developed and validated for the determination of mitiglinide in plasma using LC-MS/MS. The effect of gemfibrozil on the pharmacokinetics of orally administered mitiglinide in rats was investigated. The validated method in positive electrospray ionization
H Konya et al.
The Journal of international medical research, 37(6), 1904-1912 (2010-02-12)
This study investigated the effects of mitiglinide in 16 patients with type 2 diabetes mellitus treated with 30 mg/day mitiglinide, divided into three doses given just before each meal, for approximately 12 months. A 450 kcal meal tolerance test was
Masanori Abe et al.
Endocrine journal, 57(7), 579-586 (2010-05-15)
Mitiglinide is a rapid- and short-acting insulinotropic sulfonylurea receptor ligand and features rapid hypoglycemic action. To date, no prospective study has evaluated the use of mitiglinide in diabetic patients receiving hemodialysis (HD). In this study we evaluated the efficacy and
Haley M Phillippe et al.
The Annals of pharmacotherapy, 44(10), 1615-1623 (2010-09-16)
To review the pharmacology, pharmacokinetics, efficacy, and safety of mitiglinide, a rapid-acting insulin secretion-stimulating agent to determine its potential role in therapy for the treatment of type 2 diabetes mellitus. A MEDLINE search (1966-May 2010) was conducted for English-language, human
Jin Ah Jung et al.
Clinical drug investigation, 32(12), 799-804 (2012-10-27)
In the treatment of diabetes mellitus, combined drugs with different mechanisms of action can be effective when adequate glycaemic control is difficult with monotherapy. A fixed-dose combination (FDC) tablet of mitiglinide and metformin has been developed as a second-line treatment

Articles

We offer many products related to potassium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service