Skip to Content
Merck
All Photos(1)

Key Documents

SML0208

Sigma-Aldrich

KYP-2047

≥95% (HPLC)

Synonym(s):

(2S)-1-[[(2S)-1-(1-Oxo-4-phenylbutyl)-2-pyrrolidinyl]carbonyl]-2-pyrrolidinecarbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€158.00
25 MG
€495.00

€158.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€158.00
25 MG
€495.00

About This Item

Empirical Formula (Hill Notation):
C20H25N3O2
CAS Number:
Molecular Weight:
339.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€158.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

oil

storage condition

desiccated

color

white to light brown

storage temp.

2-8°C

SMILES string

O=C(CCCc1ccccc1)N2CCC[C@H]2C(=O)N3CCC[C@H]3C#N

InChI

1S/C20H25N3O2/c21-15-17-10-5-13-22(17)20(25)18-11-6-14-23(18)19(24)12-4-9-16-7-2-1-3-8-16/h1-3,7-8,17-18H,4-6,9-14H2/t17-,18-/m0/s1

InChI key

SPXFAUXQZWJGCJ-ROUUACIJSA-N

Application

KYP-2047 may be used in prolyl endopeptidase-mediated cell signaling studies.

Biochem/physiol Actions

KYP-2047 clears the α-synuclein aggregates induced by oxidative stress in the neuronal cells. α-synuclein is the main component of Lewy bodies in Parkinson′s disease that cause neuronal cell death and dementia.[1]
KYP-2047 is a very potent, selective inhibitor of Prolyl oligopeptidase (POP), also known as prolyl endopeptidase (PEP or PE). Ki = .023nM (porcine).
KYP-2047 is a very potent, selective inhibitor of Prolyl oligopeptidase (POP), also known as prolyl endopeptidase (PEP or PE). Ki = .023nM (porcine). KYP-2047 has been found to be neuroprotective against glutamate and oxygen, glucose deprivation in vitro. It has also been used to inhibit the formation of AcSDKP from its precursor 43-mer thymosin β4 (Tβ4). Ac-SDKP is involved in hemopoietic stem cell differentiation, is pro-angiogenic and antifibrogenic.

Features and Benefits

This compound is featured on the Neuropeptidases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T T Myöhänen et al.
British journal of pharmacology, 166(3), 1097-1113 (2012-01-12)
The aggregation of α-synuclein is connected to the pathology of Parkinson's disease and prolyl oligopeptidase (PREP) accelerates the aggregation of α-synuclein in vitro. The aim of this study was to investigate the effects of a PREP inhibitor, KYP-2047, on α-synuclein
Travis W Bainbridge et al.
Scientific reports, 7(1), 12524-12524 (2017-10-04)
Fibroblast Activation Protein (FAP) is a membrane-bound serine protease whose expression is often elevated in activated fibroblasts associated with tissue remodeling in various common diseases such as cancer, arthritis and fibrosis. Like the closely related dipeptidyl peptidase DPPIV, the extracellular
Giovanna Casili et al.
Oncotarget, 12(17), 1663-1676 (2021-08-27)
Intestinal ischemia-reperfusion (II/R) develops when the blood flow to the intestines decreases, followed by the reestablishment of the blood supply to the ischemic tissue, resulting in intestinal mucosal barrier dysfunction, with consequent severe local and systemic inflammation. Acute lung injury
Reinis Svarcbahs et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(49), 12485-12497 (2016-12-09)
Decreased clearance of α-synuclein (aSyn) and aSyn protein misfolding and aggregation are seen as major factors in the pathogenesis of Parkinson's disease (PD) and other synucleinopathies that leads to disruption in neuronal function and eventually to cell death. Prolyl oligopeptidase
Yuki Maruyama et al.
Placenta, 53, 8-15 (2017-05-11)
Prolyl oligopeptidase (prolyl endopeptidase, Prep), a multifunctional protease hydrolyzing -Pro-X- peptide bonds, is highly expressed in the mouse placenta, but the function during development is not known. We explored the possibility of Prep's involvement in placental differentiation. We cultured trophoblast

Articles

We offers many products related to neuropeptidases for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service