Skip to Content
Merck
All Photos(1)

Key Documents

SML0199

Sigma-Aldrich

Metaxalone

≥98% (HPLC)

Synonym(s):

5-(3,5-Dimethylphenoxymethyl)-2-oxazolidinone, 5-[(3,5-Xylyloxy)methyl]-2-oxazolidinone, AHR 438, Metaxalon, Metazalone, Metazolone, NSC 170959

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€53.30
50 MG
€294.00

€53.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€53.30
50 MG
€294.00

About This Item

Empirical Formula (Hill Notation):
C12H15NO3
CAS Number:
Molecular Weight:
221.25
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€53.30


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥2 mg/mL (warmed to 60 °C)

storage temp.

−20°C

SMILES string

Cc1cc(C)cc(OCC2CNC(=O)O2)c1

InChI

1S/C12H15NO3/c1-8-3-9(2)5-10(4-8)15-7-11-6-13-12(14)16-11/h3-5,11H,6-7H2,1-2H3,(H,13,14)

InChI key

IMWZZHHPURKASS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Metaxalone may be used in cell signaling studies.

Biochem/physiol Actions

Metaxalone is a muscle relaxant. The mechanism of action of metaxalone in humans is not fully understood. It was suggested that it acts through general central nervous system depression.
Metaxalone is effective to relieve pain due to acute skeletal muscle pain caused due to musculoskeletal conditions and sprains.[1][2]
Muscle relaxant (skeletal)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rakshit Kanubhai Trivedi et al.
Scientia pharmaceutica, 80(2), 353-366 (2012-08-17)
A simple, sensitive and reproducible reversed phase ultra performance liquid chromatography (RP-UPLC) coupled with a photodiode array detector method was developed for the quantitative determination of metaxalone (META) in pharmaceutical dosage forms. The method is applicable to the quantification of
Justin L Poklis et al.
Journal of analytical toxicology, 28(6), 537-541 (2004-11-02)
The case history and toxicological findings of a fatal multi-drug overdose involving metaxalone (Skelaxin) are presented. Gas-liquid chromatography with flame-ionization detection and gas chromatography-mass spectrometry were used to determine the following drug concentrations (mg/L) in aortic blood: 19 mg/L metaxalone;
Peter P Toth et al.
Clinical therapeutics, 26(9), 1355-1367 (2004-11-09)
Low back pain is a leading reason for primary care visits. Many treatment options are available, but some lack scientific support. The aim of this review was to discuss the etiology of low back pain and the relative risks and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service