Skip to Content
Merck
All Photos(1)

Key Documents

SML0079

Sigma-Aldrich

DiMNF

≥98% (HPLC)

Synonym(s):

2-(3,4-Dimethoxyphenyl)-benzo[h]chromen-4-one, NSC 123391

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16O4
CAS Number:
Molecular Weight:
332.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥13 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(cc1OC)C2=CC(=O)c3ccc4ccccc4c3O2

InChI

1S/C21H16O4/c1-23-18-10-8-14(11-20(18)24-2)19-12-17(22)16-9-7-13-5-3-4-6-15(13)21(16)25-19/h3-12H,1-2H3

InChI key

QDZQDIUUJDAORK-UHFFFAOYSA-N

Biochem/physiol Actions

DiMNF is a naphthoflavone derivative, selective aryl hydrocarbon receptor modulator (SAhRM). DiMNF is an AHR ligand that does not induce the expression of AHR dependent genes, such as CYP1A1, but inhibits the expression of cytokine-induced acute phase response proteins such as serum amyloid A and complement C3.
Selective Aryl Hydrocarbon Receptor Modulator

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark P Labrecque et al.
PloS one, 7(1), e29545-e29545 (2012-01-12)
The activated AHR/ARNT complex (AHRC) regulates the expression of target genes upon exposure to environmental contaminants such as 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD). Importantly, evidence has shown that TCDD represses estrogen receptor (ER) target gene activation through the AHRC. Our data indicates that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service