Skip to Content
Merck
All Photos(1)

Key Documents

SMB01026

Sigma-Aldrich

Mosloflavon

≥90% (LC/MS-ELSD)

Synonym(s):

5-Hydroxy-6,7-dimethoxyflavone

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€183.00

€183.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€183.00

About This Item

Empirical Formula (Hill Notation):
C17H14O5
CAS Number:
Molecular Weight:
298.29
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

€183.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

298.29

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

[o]1c2c([c](cc1c3ccccc3)=O)c(c(c(c2)OC)OC)O

InChI

1S/C17H14O5/c1-20-14-9-13-15(16(19)17(14)21-2)11(18)8-12(22-13)10-6-4-3-5-7-10/h3-9,19H,1-2H3

InChI key

SIVAITYPYQQYAP-UHFFFAOYSA-N

General description

Mosloflavon, a 2-hydroxyflavanone, is a bioactive natural compound commonly derived from plants such as Mosla soochouensis, Desmos dumosus, Muntingia calabura, Cryptocarya chinensis, and Phonus arborescens. Extensive research indicates that this natural metabolite acts as an inhibitor may possess a wide range of biological activities, including anti-inflammatory, antioxidant, antiviral, antifungal, immunomodulatory, anti-infection, and anti-hypoxic properties.

Application

Mosloflavon is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Mosloflavanone exhibits potent anti-EV71 activity by reducing visible cytopathic effects and suppressing virus replication during the initial infection phase. It also hinders the expression of the viral VP2 protein, thereby impeding viral capsid protein synthesis. Additionally, Mosloflavone demonstrates significant anti-inflammatory potential through the inhibition of TNF-α and IL-1β, with IC50 values of 16.4 μM and 6.4 μM, respectively. It shows a dose-dependent reduction in TNF-α, IL-1β, and iNOS levels in the supernatant of the mouse macrophage cell line J774A. This compound holds promise as a foundational element in the quest for treatments targeting inflammatory and immunomodulatory diseases.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service