Skip to Content
Merck
All Photos(1)

Documents

SMB00182

Sigma-Aldrich

Eudesmine

≥95% (LC/MS-ELSD)

Synonym(s):

Eudesamin, Eudesmin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26O6
CAS Number:
Molecular Weight:
386.44
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1ccc(cc1OC)[C@@H]2OCC3C2CO[C@H]3c4ccc(OC)c(OC)c4

InChI

1S/C22H26O6/c1-23-17-7-5-13(9-19(17)25-3)21-15-11-28-22(16(15)12-27-21)14-6-8-18(24-2)20(10-14)26-4/h5-10,15-16,21-22H,11-12H2,1-4H3/t15-,16-,21+,22+/m1/s1

InChI key

PEUUVVGQIVMSAW-DJDZNOHASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Biochem/physiol Actions

Eudesmin is a component of a Brazilian folk medicine that has been used for blood pressure reduction. Eudesmin also is an inhibitor of TNF-alpha production and T cell proliferation.
Found in the Rutaceae family that has estrogenic and sedative effects.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Subhas Chandra Roy et al.
The Journal of organic chemistry, 67(10), 3242-3248 (2002-05-11)
Intramolecular radical cyclization of suitably substituted epoxy ethers 4a-g using bis(cyclopentadienyl)titanium(III) chloride as the radical source resulted in trisubstituted tetrahydrofurano lignans and 2,6-diaryl-3,7-dioxabicyclo[3.3.0]octane lignans depending on the reaction conditions. The titanium(III) species was prepared in situ from commercially available titanocene
Juliana Montani Raimundo et al.
European journal of pharmacology, 606(1-3), 150-154 (2009-04-21)
In Brazilian folk medicine, extracts from Piper species are used to reduce blood pressure. Previously, we demonstrated the vasodilatory activity of crude extracts from leaves of Piper truncatum explaining their possible use in the treatment of hypertension in traditional medicine.
Isabele R Nascimento et al.
Pest management science, 60(4), 413-416 (2004-05-04)
Acetone and ethanol extracts of the tubercula and several compounds isolated from Aristolochia pubescens (Willd) were bioassayed on velvetbean caterpillars, Anticarsia gemmatalis (Hübner), for evaluation of the insecticidal activities. Of the extracts subjected to bioassay, the acetone extract showed the
Yoo Jung Yang et al.
Archives of pharmacal research, 29(12), 1114-1118 (2007-01-18)
(+)-Eudesmin [4,8-bis(3,4-dimethoxyphenyl)-3,7-dioxabicyclo[3.3.0]octane] was isolated from the stem bark of Magnolia kobus DC. var. borealis Sarg. and found to have neuritogenic activity. 50 microM (+)-eudesmin induced neurite outgrowth and enhanced nerve growth factor (NGF)-mediated neurite outgrowth from PC12 cells. At this
J Y Cho et al.
Archives of pharmacal research, 22(4), 348-353 (1999-09-18)
Possible antiinflammatory effects of eudesmin were examined by assessing the effects on tumor necrosis factor (TNF)-alpha production and lymphocyte proliferation as well as cytotoxicity against murine and human macrophages. The compound significantly inhibited TNF-alpha production by lipopolysaccharide (LPS)-stimulated murine macrophage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service