Skip to Content
Merck
All Photos(1)

Key Documents

SMB00144

Sigma-Aldrich

Labdanolic acid

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€305.00

€305.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€305.00

About This Item

Empirical Formula (Hill Notation):
C20H36O3
CAS Number:
Molecular Weight:
324.50
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

€305.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C[C@@H](CC[C@H]1[C@](C)(O)CC[C@H]2C(C)(C)CCC[C@]12C)CC(O)=O

InChI

1S/C20H36O3/c1-14(13-17(21)22)7-8-16-19(4)11-6-10-18(2,3)15(19)9-12-20(16,5)23/h14-16,23H,6-13H2,1-5H3,(H,21,22)/t14-,15-,16+,19-,20+/m0/s1

InChI key

KHCCSRVJJDOANA-RQOBALISSA-N

General description

Labdanolic acid is a key constituent of the acid fraction of Cistus ladaniferus. It is an easily isolable natural product.[1]

Biochem/physiol Actions

Labdanolic acid acts as a synthetic precursor for active drimanes.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Labdanolic Acid as Synthetic Precursor of Active Drimanes
Lithgow A M, et al.
Natural Product Letters, 6(4), 291-294 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service