Skip to Content
Merck
All Photos(1)

Documents

SMB00093

Sigma-Aldrich

Pipernonaline

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27NO3
CAS Number:
Molecular Weight:
341.44
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O=C(\C=C\CCCC\C=C\c1ccc2OCOc2c1)N3CCCCC3

InChI

1S/C21H27NO3/c23-21(22-14-8-5-9-15-22)11-7-4-2-1-3-6-10-18-12-13-19-20(16-18)25-17-24-19/h6-7,10-13,16H,1-5,8-9,14-15,17H2/b10-6+,11-7+

InChI key

PKLGRWSJBLGIBF-JMQWPVDRSA-N

General description

Natural product derived from plant source.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuang Jin et al.
Phytotherapy research : PTR, 23(8), 1194-1196 (2009-01-28)
A bioassay-guided isolation of an ethanol extract of the fruit of Piper longum L. yielded piperlonguminine, piperine and pipernonaline, as the main antihyperlipidemic constituents. They exhibit appreciable antihyperlipidemic activity in vivo, which is comparable to that of the commercial antihyperlipidemic
Seung Woong Lee et al.
Bioorganic & medicinal chemistry letters, 18(16), 4544-4546 (2008-08-02)
Eight alkamides 1-8 were isolated by bioassay-guided isolation of EtOH extracts of the fruits of Piper longum and Piper nigum (Piperaceae). Their structures were elucidated by spectroscopic analysis ((1)H, (13)C NMR, and ESI-MS) as follows: guineensine (1), retrofracamide C (2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service