Skip to Content
Merck
All Photos(1)

Key Documents

SIC008

Sigma-Aldrich

MISSION® siRNA Fluorescent Universal Negative Control #2, 6-FAM

Synonym(s):

MISSION® siRNA Fluorescent Universal Negative Control, 6-FAM

Sign Into View Organizational & Contract Pricing

Select a Size

1 NMOL
€124.00
10 NMOL
€256.00

€124.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 NMOL
€124.00
10 NMOL
€256.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€124.00


Please contact Customer Service for Availability

Request a Bulk Order

product line

MISSION®

color

Green (6-FAM)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Negative controls are an essential part of every siRNA experiment, and here we offer the siRNA Universal Negative Control #2 conjugated to 6-FAM for easy visualization and assessment of transfection efficiency.
Negative controls are an essential part of every siRNA experiment, and here we offer the siRNA Universal Negative Control #2 conjugated to Cyanine 3 for easy visualization and assessment of transfection efficiency.

Features and Benefits

  • 6-FAM labeled siRNA Universal Negative Control allows easy visualization of the siRNA in your cells by using fluorescence microscopy.
  • Rapidly assess transfection efficiency.
  • Ready to use.
  • Available in three sizes for your convenience.
  • Compatible with Human, Mouse, and Rat cells.

Legal Information

All siRNA oligos are sold under license from the Massachusetts Institute of Technology.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

MISSION siRNA Negative Controls ensure mRNA knockdown efficiency baseline; tested in human, rat, and mouse cells, designed without gene homology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service