Skip to Content
Merck
All Photos(1)

Key Documents

SHH0022

Sigma-Aldrich

Histoheme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500

application(s)

hematology
histology

storage temp.

room temp

General description

Developed to assist histologists with bloody specimens such as placental fragments, bone marrow biopsies, liver, spleen and endometrial curettings which tend to harden after processing and dry out. When set on ice, these tissues are prone to shatter and flake, leaving blood fragments on the work area, microtome, and water bath, and have a high potential to create extraneous tissue “floaters” on slides.

Histoheme is an ammonia based emollient that alleviates these problems and mess. Trimming into these blocks and soaking tissue in Histoheme allows technicians to cut clean, non-fragmented sections without stopping in-between each block to allow for better morphological staining.

Testing Method

Procedure
1. Trim blocks 10-20 microns.
2. Soak in small amounts of Histoheme. After use, dispose of used Histoheme to prevent cross-contamination with other samples.
3. Place block in microtome chuck and recut for 4-5 μm sections.

Histoheme does not have any effect on H&E staining or immunohistochemistry stains. The product is completely removed during the deparaffinization process.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service