Skip to Content
Merck
All Photos(1)

Key Documents

SCP0250

Sigma-Aldrich

Syntide 2 Peptide

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€115.00
5 MG
€399.00

€115.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
€115.00
5 MG
€399.00

About This Item

Empirical Formula (Hill Notation):
C68H122N20O18
Molecular Weight:
1507.82
UNSPSC Code:
12352204
NACRES:
NA.32

€115.00


Please contact Customer Service for Availability

Product Name

Syntide 2,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sudhir Kumar et al.
mBio, 12(6), e0257521-e0257521 (2021-11-03)
Gametocytes of the malaria parasite Plasmodium are taken up by the mosquito vector with an infectious blood meal, representing a critical stage for parasite transmission. Calcium-independent protein kinases (CDPKs) play key roles in calcium-mediated signaling across the complex life cycle
Lushuang Huang et al.
Analytical chemistry, 92(1), 640-646 (2019-12-04)
The analysis of low-volume samples provides valuable insight into complex biological systems. However, the proteomic and metabolomic analysis of low-volume samples remains challenging due to the lack of simple, efficient, and reproducible microsampling techniques. We have developed an electrospray-assisted device
Nathália Rocco-Machado et al.
The Journal of biological chemistry, 298(11), 102579-102579 (2022-10-12)
Calcium/calmodulin-dependent protein kinase II δ (CaMKIIδ) has a pivotal role in cardiac signaling. Constitutive and deleterious CaMKII "autonomous" activation is induced by oxidative stress, and the previously reported mechanism involves oxidation of methionine residues in the regulatory domain. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service