Skip to Content
Merck
All Photos(1)

Key Documents

SCP0079

Sigma-Aldrich

Caspase 2/3 Inhibitor

≥94% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H37N5O13
Molecular Weight:
603.58
UNSPSC Code:
12352200
Product SCP0079 is not currently sold in your country. Contact Technical Service

Assay

≥94% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Leu-Asp-Glu-Ser-Asp-al

Application

Caspase 2/3 Inhibitor (Ac-LDESD-CHO) is used to study the active site recognition of penta5 (P5) position substrates by caspases 2 and 3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Schweizer et al.
The Journal of biological chemistry, 278(43), 42441-42447 (2003-08-16)
The cell death protease caspase-2 has recently been recognized as the most apical caspase in the apoptotic cascade ignited during cell stress signaling. Cytotoxic stress, such as that caused by cancer therapies, leads to activation of caspase-2, which acts as
Guoxing Fu et al.
Apoptosis : an international journal on programmed cell death, 13(11), 1291-1302 (2008-09-10)
Caspase-3, -6 and -7 cleave many proteins at specific sites to induce apoptosis. Their recognition of the P5 position in substrates has been investigated by kinetics, modeling and crystallography. Caspase-3 and -6 recognize P5 in pentapeptides as shown by enzyme
Ling Wang et al.
Cancer research, 68(12), 4640-4648 (2008-06-19)
Kaposi's sarcoma-associated herpesvirus (KSHV) is associated with three different human malignancies, including Kaposi's sarcoma (KS), primary effusion lymphoma, and multicentric Castleman's disease. The KS lesion is of endothelial cell in origin and is highly dependent on autocrine and paracrine factors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service