Skip to Content
Merck
All Photos(1)

Key Documents

SCP0051

Sigma-Aldrich

[Glp3]-Amyloid β 3-42 human

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€526.00

€526.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
€526.00

About This Item

Empirical Formula (Hill Notation):
C196H299N53O55S1
Molecular Weight:
4309.86
UNSPSC Code:
12352209
NACRES:
NA.32

€526.00


Please contact Customer Service for Availability

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Glp-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala

Application

Amyloid β (Aβ) refers to peptides derived from Amyloid precursor protein that vary in length from 36-43 amino acids. Aβ(s) peptides, their peptide fragments and mutated fragments are used to study a wide range of metabolic and regulatory functions including activation of kinases, regulation of cholesterol transport, function as a transcription factor, and regulators of inflammation. Aβ(s) peptides and their peptide fragments are also used to study oxidative stress, metal binding and mechanisms of protein cross-linking in the context of diseases such as Alzheimer′s disease and neurodegeneration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Wirths et al.
Acta neuropathologica, 118(4), 487-496 (2009-06-24)
It is well established that only a fraction of Abeta peptides in the brain of Alzheimer's disease (AD) patients start with N-terminal aspartate (Abeta(1D)) which is generated by proteolytic processing of amyloid precursor protein (APP) by BACE. N-terminally truncated and
Gawain McColl et al.
The Journal of biological chemistry, 284(34), 22697-22702 (2009-07-04)
Transgenic expression of human amyloid beta (A beta) peptide in body wall muscle cells of Caenorhabditis elegans has been used to better understand aspects of Alzheimer disease (AD). In human aging and AD, A beta undergoes post-translational changes including covalent
Heinke Schieb et al.
The Journal of biological chemistry, 286(39), 33747-33758 (2011-07-29)
In this study, we report a detailed analysis of the different variants of amyloid-β (Aβ) peptides in the brains and the cerebrospinal fluid from APP23 transgenic mice, expressing amyloid precursor protein with the Swedish familial Alzheimer disease mutation, at different

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service