Skip to Content
Merck
All Photos(1)

Documents

SCP0018

Sigma-Aldrich

[Asn1,Val5,Asn9]-Angiotensin I salmon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H87N17O14
Molecular Weight:
1258.43
UNSPSC Code:
12352209
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Asn-Arg-Val-Tyr-Val-His-Pro-Phe-Asn-Leu

Application

Angiotensins are peptide hormone components of the renin-angiotensin system involved in the regulation of blood vessel constriction and aldosterone release. Angiotensin I, a decapeptide derived from angiotensinogen, is a converted to Angiotensin II by the removal of the two C-terminal amino acids. Various analogues of Angiotensin I may be used as substrates to identify, differentiate and characterize angiotensin-converting enzyme(s) (ACE).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshio Takei et al.
General and comparative endocrinology, 135(3), 286-292 (2004-01-16)
In order to delineate further the molecular evolution of the renin-angiotensin system in vertebrates, angiotensin I (ANG I) has been isolated after incubation of plasma and kidney extracts of emu (Dromiceus novaehollandiae), axolotl (Ambystoma mexicanum), and sea lamprey (Petromyzon marinus).
M C Khosla et al.
Hypertension (Dallas, Tex. : 1979), 5(6 Pt 3), V22-V28 (1983-11-01)
To understand how vertebrates utilize angiotensins during evolutionary development, we undertook studies to synthesize and/or characterize angiotensin-like peptides from nonmammalian species. The present paper describes the synthesis of [Asp1,Val5,Asn9] angiotensin I (bull frog, Rana catesbeiana) (I), [Asn1,Val5,His9] angiotensin I (Japanese
N J Bernier et al.
The American journal of physiology, 273(1 Pt 2), R49-R57 (1997-07-01)
Immunohistochemical and pharmacological techniques were utilized to investigate the relationships between angiotensins and catecholamine release from the chromaffin tissue of rainbow trout (Oncorhynchus mykiss). Double labeling with [Asp1, Ile5]angiotensin II-fluorescein isothiocyanate (ANG II-FITC) and anti-dopamine beta-hydroxylase revealed specific ANG II

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service