SCJ4600018
CF™ 488A succinimidyl ester
suitable for fluorescence
Select a Size
€304.00
Select a Size
About This Item
€304.00
Recommended Products
form
solid
Quality Level
mol wt
~914 g/mol
ε (extinction coefficient)
70000 at 490 nm
absorbance ratio
A280/490 nm 0.1
fluorescence
λex 488 nm (Flow cytometry laser line)
λex 488 nm (Microscopy laser line)
λex 490 nm; λem 515 nm
suitability
suitable for fluorescence
shipped in
wet ice
storage temp.
−20°C
General description
Preparation Note
Legal Information
Disclaimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service