Skip to Content
Merck
All Photos(1)

Key Documents

SBR00057

Sigma-Aldrich

Retapamulin

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€129.00

€129.00


Please contact Customer Service for Availability


Select a Size

Change View
50 MG
€129.00

About This Item

Empirical Formula (Hill Notation):
C30H47NO4S
CAS Number:
Molecular Weight:
517.76
MDL number:
UNSPSC Code:
51284706
NACRES:
NA.76

€129.00


Please contact Customer Service for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

S(C4C[C@@H]5N([C@@H](CC5)C4)C)CC(=O)O[C@H]1[C@]2([C@H]3[C@@]([C@H]([C@@H]([C@](C1)(C)C=C)O)C)(CC[C@H]2C)CCC3=O)C

InChI

1S/C30H47NO4S/c1-7-28(4)16-24(35-25(33)17-36-22-14-20-8-9-21(15-22)31(20)6)29(5)18(2)10-12-30(19(3)27(28)34)13-11-23(32)26(29)30/h7,18-22,24,26-27,34H,1,8-17H2,2-6H3/t18-,19+,20-,21+,22?,24-,26+,27+,28-,29+,30+/m1/s1

InChI key

STZYTFJPGGDRJD-FJJJPKKESA-N

General description

Retapamulin (RET) is a semi-synthetic antibiotic ointment, and pleuromutilin derivative compound.[1] The primary structural difference between pleuromutilin and retapamulin is in the C-14 side chain.[1]

Biochem/physiol Actions

Retapamulin is a potent antibacterial agent against Gram-positive bacteria, especially against Staphylococcus aureus and S. pyogenes.[1] It displays synergistic effects against Enterococcus faecalis in combination with streptogramin (MLS).[2] RET initiates ribosomal arrests at the start codon and enables genes to use alternative translational sites in E. coli.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro synergistic effect of retapamulin with erythromycin and quinupristin against Enterococcus faecalis
Park B, et al
The Journal of Antibiotics, 73, 630-635 (2020)
Retapamulin: Current Status and Future Perspectives
Goudarzi M, et al
Archives of Clinical Infectious Diseases, 16, 4-4 (2021)
Retapamulin-Assisted Ribosome Profiling Reveals the Alternative Bacterial Proteome
Meydan S, et al
Molecular Cell, 74, 481-493 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service