Skip to Content
Merck
All Photos(1)

Documents

SAE0066

Sigma-Aldrich

Adenylyl Cyclase Toxin from Bordetella pertussis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

Bordetella pertussis Tohama I

Quality Level

Assay

≥70%

form

liquid

specific activity

≥ 50  units/mg protein

technique(s)

cell culture | mammalian: suitable

suitability

suitable for molecular biology

application(s)

detection

storage temp.

−20°C

Gene Information

Bordetella pertussis Tohama I ... CyaA(69600712)

General description

Research area: IMMUNO AND CKS

Adenylate Cyclase Toxin (ACT or CyaA) is a member of the extensive family of toxins known as Repeat in Toxin (RTX), which are produced by Gram-negative organisms. ACT is encoded by the cyaA gene and secreted extracellularly in the form of a soluble protein. It exhibits both adenylate cyclase enzymatic activity and hemolytic activity. The synthesis, maturation, and secretion of ACT are regulated by the CyaCABD operon. Moreover, its specific cellular receptor, CD11b/CD18 integrin (αMβ2, Mac-1, or CR3), is expressed on myeloid phagocytes.

Application

Adenylyl Cyclase Toxin from Bordetella pertussis has been used as Gα(i/o) inhibitor to study the involvement of the sphingosine 1-phosphate receptor 2/Gα(12/13)/MAPK signaling pathway in the priming and activation of NLRP3 inflammasome during cholestatic liver injury.

Biochem/physiol Actions

Adenylate Cyclase Toxin (CyaA) is responsible for inhibiting the phagocytic activities of neutrophils and macrophages by impairing oxidative response and chemotaxis, ultimately leading to cell apoptosis or necrosis. Additionally, ACT can upregulate the expression of MHC class II and costimulatory molecules on dendritic cells, thereby reducing proinflammatory cytokine production.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pertussis toxin and adenylate cyclase toxin: key virulence factors of Bordetella pertussis and cell biology tools
Carbonetti NH
Future Microbiology, 5, 455?469-455?469 (2010)
Understanding the Mechanism of Translocation of Adenylate Cyclase Toxin across Biological Membranes
Ostolaza H, et al.
Toxins, 9(10), 295-295 (2017)
Bioengineering of Bordetella pertussis Adenylate Cyclase Toxin for Antigen-Delivery and Immunotherapy
Chenal A and Ladant D
Toxins, 10(7), 302-302 (2018)
NLRP3 inflammasome priming and activation in cholestatic liver injury via the sphingosine 1-phosphate/S1P receptor 2/G?(12/13)/MAPK signaling pathway
Hou L, et al.
Journal of Molecular Medicine, 99, 273?288-273?288 (2021)
Lei Hou et al.
Journal of molecular medicine (Berlin, Germany), 99(2), 273-288 (2021-01-04)
NLRP3 inflammasome-driven inflammation represents a key trigger for hepatic fibrogenesis during cholestatic liver injury. However, whether sphingosine 1-phosphate (S1P) plays a role in NLRP3 inflammasome priming and activation remains unknown. Here, we found that the expression of NLRP3 in macrophages

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service