Skip to Content
Merck
All Photos(1)

Key Documents

SAB5900401

Sigma-Aldrich

Anti-Monkeypox virus/MPXV H3L Antibody (SAA0282)

new

Synonym(s):

H3L, Monkeypox virus (MPXV)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
Pricing and availability is not currently available.

biological source

mouse

Quality Level

recombinant

expressed in CHO cells

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

clone

SAA0282, recombinant monoclonal

Assay

>95% (SDS-PAGE)

form

liquid

mol wt

150 kDa

species reactivity

monkeypox virus

concentration

1 mg/mL

technique(s)

ELISA: 1:2000-1:4000
neutralization: 0.01-10 μg/mL

color

colorless

pH

7.4

isotype

IgG2a, kappa

Protein ID accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

-65 to -96°C

target post-translational modification

unmodified

General description

Anti-Monkeypox virus/MPXV H3L Antibody, clone SAA0282 is produced in mouse. The monoclonal antibodies are IgG immunoglobulins, which consist of two heavy chains and two light chains. Each heavy chain contains four domains, and each light chain contains two domains. The antibody binds to the antigen of the monkeypox virus, which is the H3L protein.




Monkeypox virus (MPXV), like smallpox virus, belongs to the genus Orthopoxvirus. Its morphology is consistent with that of orthopoxvirus, with a brick-shaped or oval shape with rounded corners and a size of 200~400nm. Brick-shaped or oval-shaped, covered by a lipoprotein membrane or envelope, with two protein-containing side bodies in the middle, and a thick membrane core containing a large double-stranded DNA gene body. The genome of Mpox virus is a double-stranded DNA, about 197kb long, and the end of the genome contains an identical but opposite terminal inverted repeat sequence. The virus contains 190 open reading frames, 4 of which are located in terminal inverted repeats. The core proteins of monkeypox virus are A29L, A35R, B6R, M1R, H3L.

Specificity

Monoclonal Anti-Monkeypox virus/MPXV H3L Antibody, clone SAA0282 is highly specific and bind to the H3L protein of the virus. This binding inhibits the virus from entering cells, thus preventing infection. The antibodies also trigger the body’s immune system to produce anti-viral proteins that can clear the virus from the body.

Application

ELISA,neutralization

Packaging

100µg per vial / 1 mg per vial

Storage and Stability

Use a manual defrost freezer and avoid repeated freeze-thaw cycles.Store at +4°C short term (1-2 weeks).Store at -20 °C 12 months. Store at -80°C long term.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service