Skip to Content
Merck
All Photos(2)

Key Documents

SAB5801058

Sigma-Aldrich

Goat Anti-Rabbit IgG(Fcγ) Alpaca Heavy chain only Antibody

recombinant Alpaca(VHH) Nano antibody

Synonym(s):

Recombinant Alpaca single variable domain secondary Nanobody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€289.00

€289.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€289.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€289.00


Please contact Customer Service for Availability

biological source

alpaca

recombinant

expressed in HEK 293 cells (Human cell line)

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

recombinant monoclonal

form

lyophilized solid

mol wt

100 kDa

species reactivity

Alpaca, rabbit

technique(s)

ELISA: 1:10000-1:50000
western blot: 1:10000-1:50000

size

0.1 mL

color

colorless

isotype

VHH

shipped in

wet ice

storage temp.

2-8°C

General description

Goat Anti-Rabbit IgG(Fcγ) Alpaca Heavy chain only Antibody is designed to specifically detect Fc region of rabbit IgG Fcγ fragment. It is a monovalent, single domain, recombinant antibody fused to goat IgG Fc. Based on immunoelectrophoresis and/or ELISA, Goat Anti-Rabbit IgG(Fcγ) Alpaca Heavy chain only Antibody reacts with the Fc region of rabbit IgG selectively but not with mouse, human, cynomolgus, rat, goat IgG.
Rabbit research antibodies are widely used in life science research. So far, four isotypes have been identified (IgA, IgE, IgG, and IgM) in rabbits. Each isotype has a different heavy chain. Rabbit has only one IgG subclass. The whole IgG molecule possesses both the Fc region and the Fab region, which possessing the epitope-recognition site. The IgG contains two heavy and light chains. The heavy chain is about 50 KD and the light chain is about 25 KD. The common IgG is monomeric with a molecular weight of approximately 150 kD.
VHH are single-domain antibodies derived from the variable regions of heavy chain of Camelidae immunoglobulin. The size of VHH is extremely small(less than15KDa) compared to other forms of antibody fragment, which significantly increase the permeability of VHH. Thus VHH is considered of great value for research, diagnostics and therapeutics.

Specificity

Fc region of Rabbit IgG
Isotype: VHH domain of alpaca IgG2b/2c fused to goat IgG Fc

Immunogen

Recombinant Alpaca single variable domain secondary nano antibody

Packaging

100 μg in Glass bottle

Physical form

Lyophilized from 10mM PBS (pH 7.5), 0.05% sucrose, 0.1% trehalose

Reconstitution

Add 100uL dH2O or 100uL dH2O with 50% Glycerol to a final concentration of 1mg/mL

Storage and Stability

After reconstitution, store at –20 °C(Avoid freeze / thaw cycles). Stable for 12 months at -20°C

Other Notes

No cross-reactivity with mouse, human, cynomolgus, rat, goat IgG

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service