Skip to Content
Merck
All Photos(5)

Key Documents

SAB5702948

Sigma-Aldrich

Anti-E-Cadherin Antibody, clone 2Q9Y7, Rabbit Monoclonal

Synonym(s):

Arc-1,CD324,CDHE,ECAD,LCAM,UVO,CDH1,E-Cadherin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€377.00

€377.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€377.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€377.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

2Q9Y7, monoclonal

form

liquid

mol wt

135 kDa

species reactivity

human, rat, rat

concentration

1.00 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:1000

color

colorless

isotype

IgG

immunogen sequence

PVEAGLQIPAILGILGGILALLILILLLLLFLRRRAVVKEPLLPPEDDTRDNVYYYDEEGGGEEDQDFDLSQLHRGLDARPEVTRNDVAPTLMSVPRYLPR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... CDH1(999)

General description

This gene encodes a classical cadherin of the cadherin superfamily. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate the mature glycoprotein. This calcium-dependent cell-cell adhesion protein is comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function of this gene is thought to contribute to cancer progression by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. This gene is present in a gene cluster with other members of the cadherin family on chromosome 16.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 700-800 of human E-Cadherin (NP_004351.1).
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC, IF/ICC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service