Skip to Content
Merck
All Photos(3)

Key Documents

SAB5702811

Sigma-Aldrich

Anti-IGHD Antibody, clone 5M8U3, Rabbit Monoclonal

Synonym(s):

immunoglobulin heavy constant delta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€377.00

€377.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€377.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€377.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

5M8U3, monoclonal

form

liquid

mol wt

55 kDa

species reactivity

human

concentration

1.1 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

APTKAPDVFPIISGCRHPKDNSPVVLACLITGYHPTSVTVTWYMGTQSQPQRTFPEIQRRDSYYMTSSQLSTPLQQWRQGEYKCVVQHTASKSKKEIFRW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGHD(3495)

General description

Constant region of immunoglobulin heavy chains. Immunoglobulins, also known as antibodies, are membrane-bound or secreted glycoproteins produced by B lymphocytes. In the recognition phase of humoral immunity, the membrane-bound immunoglobulins serve as receptors which, upon binding of a specific antigen, trigger the clonal expansion and differentiation of B lymphocytes into immunoglobulins-secreting plasma cells. Secreted immunoglobulins mediate the effector phase of humoral immunity, which results in the elimination of bound antigens.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human IGHD (P01880).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service