Skip to Content
Merck
All Photos(4)

Key Documents

SAB5702738

Sigma-Aldrich

Anti-LIN28A Antibody, clone 2S1I10, Rabbit Monoclonal

Synonym(s):

CSDD1, LIN-28, LIN28, ZCCHC1, lin-28A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€377.00

€377.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€377.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€377.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

2S1I10, monoclonal

form

liquid

mol wt

26 kDa

species reactivity

human, rat, rat

concentration

0.70 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:10000 - 1:20000

color

colorless

isotype

IgG

immunogen sequence

MGSVSNQQFAGGCAKAAEEAPEEAPEDAARAADEPQLLHGAGICKWFNVRMGFGFLSMTARAGVALDPPVDVFVHQSKLHMEGFRSLKEGEAVEFTFKKS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIN28A(79727)

General description

This gene encodes a LIN-28 family RNA-binding protein that acts as a posttranscriptional regulator of genes involved in developmental timing and self-renewal in embryonic stem cells. The encoded protein functions through direct interaction with target mRNAs and by disrupting the maturation of certain miRNAs involved in embryonic development. This protein prevents the terminal processing of the LET7 family of microRNAs which are major regulators of cellular growth and differentiation. Aberrant expression of this gene is associated with cancer progression in multiple tissues. [provided by RefSeq, Sep 2015]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LIN28A (Q9H9Z2).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service