Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702219

Sigma-Aldrich

Anti-HSPA12A Antibody, clone 8Q8M3, Rabbit Monoclonal

Synonym(s):

heat shock 70kDa protein 12A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€377.00

€377.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€377.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€377.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

material

colorless

clone

8Q8M3, monoclonal

form

liquid

mol wt

75 kDa

species reactivity

human, rat, rat

concentration

1.50 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

SEQQSFLVVVAVDFGTTSSGYAYSFTKEPECIHVMRRWEGGDPGVSNQKTPTTILLTPERKFHSFGYAARDFYHDLDPNEAKQWLYLEKFKMKLHTTGDLT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Adapter protein for SORL1, but not SORT1. Delays SORL1 internalization and affects SORL1 subcellular localization.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 50-150 of human HSPA12A (O43301).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service