Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702048

Sigma-Aldrich

Anti-DNA Polymerase gamma Antibody, clone 9V9D9, Rabbit Monoclonal

Synonym(s):

MDP1, MIRAS, MTDPS4A, MTDPS4B, PEO, POLG1, POLGA, SANDO, SCAE

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€377.00

€377.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€377.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€377.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

9V9D9, monoclonal

form

liquid

mol wt

140 kDa

species reactivity

rat, human, rat

concentration

1 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

LVREEDRYRAALALQITNLLTRCMFAYKLGLNDLPQSVAFFSAVDIDRCLRKEVTMDCKTPSNPTGMERRYGIPQGEALDIYQIIELTKGSLEKRSQPGP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POLG(5428)

General description

Mitochondrial DNA polymerase is heterotrimeric, consisting of a homodimer of accessory subunits plus a catalytic subunit. The protein encoded by this gene is the catalytic subunit of mitochondrial DNA polymerase. The encoded protein contains a polyglutamine tract near its N-terminus that may be polymorphic. Defects in this gene are a cause of progressive external ophthalmoplegia with mitochondrial DNA deletions 1 (PEOA1), sensory ataxic neuropathy dysarthria and ophthalmoparesis (SANDO), Alpers-Huttenlocher syndrome (AHS), and mitochondrial neurogastrointestinal encephalopathy syndrome (MNGIE). Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1140-1239 of human DNA Polymerase gamma (P54098).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service