Skip to Content
Merck
All Photos(3)

Key Documents

SAB5701471

Sigma-Aldrich

Anti- RB antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€357.00

€357.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€357.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
monoclonal
application:
IHC
WB
species reactivity:
human, mouse, rat
technique(s):
immunohistochemistry: 1:50-1:200
western blot: 1:500-1:2000
citations:

€357.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

monoclonal

form

liquid

species reactivity

human, mouse, rat

concentration

1.5 mg/mL

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RB1(5925)

General description

The protein encoded by this gene is a negative regulator of the cell cycle and was the first tumor suppressor gene found. The encoded protein also stabilizes constitutive heterochromatin to maintain the overall chromatin structure. The active, hypophosphorylated form of the protein binds transcription factor E2F1. Defects in this gene are a cause of childhood cancer retinoblastoma (RB), bladder cancer, and osteogenic sarcoma. [provided by RefSeq, Jul 2008]

Immunogen

A synthesized peptide derived from human RB

Physical form

Buffer: PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service