Skip to Content
Merck
All Photos(2)

Key Documents

SAB5700595

Sigma-Aldrich

Recombinant 2019-nCoV envelope Protein with His and Avi tag

Synonym(s):

2019-nCoV E protein, 2019-nCoV sM protein, Env polyprotein, Envelope glycoprotein, Envelope protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€389.00

€389.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€389.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€389.00


Please contact Customer Service for Availability

recombinant

expressed in E. coli

Quality Level

Assay

>95% (SDS-PAGE)

form

liquid

mol wt

11.4 kDa

impurities

<0.1 EU/μg protein (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

SARS virus ... E(43740570)

General description

Recombinant SARS-COV-2(2019-nCoV) envelope Protein is produced by E. coli expression system. The target protein is expressed with sequence (Met1-Val75) of SARS-COV-2(2019-nCoV) envelope (Accession #QHD43418.1) fused with an initial Met, a 6XHis, Avi tag at the N-terminus.

Biochem/physiol Actions

Measured by its binding ability in a functional ELISA. Immobilized Recombinant 2019-nCoV Envelope at 2μg/mL (100 μL/well) can bind Recombinant 2019-nCoV Nucleocapsid with a linear range of 1.2-41.1 ng/mL.

Storage and Stability

Store the lyophilized protein at -20°C to -80°C for long term. After reconstitution, the protein solution is stable at -20°C for 3 months, at 2-8 °C for up to 1 week. This product is stable at ≤ -70°C for up to 6 months from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Data presented is the available current product information and provided as-is. These products have not been tested or verified in any additional applications, sample types, including any clinical use. Experimental conditions must be empirically derived by the user. Our Antibody Guarantee only covers tested applications stated herein and conditions presented in our product information and is not extended to publications.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service