Skip to Content
Merck
All Photos(4)

Key Documents

SAB5700593

Sigma-Aldrich

Recombinant 2019-nCoV Spike S1 Protein with His and Avi tag

Synonym(s):

S glycoprotein Subunit1, S1 protein, Spike glycoprotein Subunit1, Spike protein S1, novel coronavirus s1 Protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€389.00

€389.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€389.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€389.00


Please contact Customer Service for Availability

recombinant

expressed in HEK 293 cells

Quality Level

Assay

>95% (HPLC)
>95% (SDS-PAGE)

form

lyophilized powder

impurities

<1.0 EU/μg protein (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

SARS virus ... S(43740568)

General description

The spike protein (S) of coronavirus (CoV) attaches the virus to its cellular receptor, angiotensin-converting enzyme 2 (ACE2). A defined receptor-binding domain (RBD) on S mediates thisinteraction.The S protein plays key parts in the induction of neutralizing-antibody and T-cellresponses, as well as protective immunity.
Recombinant SARS-COV-2(2019-nCoV) Spike S1 Protein is produced by HEK293 cells expression system. The target protein is expressed with sequence (Gln14-Arg683) of SARS-COV-2(2019-nCoV) Spike S1 (Accession #YP_009724390.1) fused with a 6XHis tag and Avi at the C-terminus.

Biochem/physiol Actions

Measured by its binding ability in a functional ELISA. Immobilized Recombinant SARS-CoV-2 Spike S1 at 2μg/mL (100 μL/well) can bind recombinant Human ACE2 with a linear range of 0.15-6.85 ng/mL.|Immobilized Human ACE2 on COOH Chip, can bind SARS-COV-2 Spike S1 with an affinity constant of 24.5 nM as determined in a SPR assay.

Reconstitution

For lyophilized protein : Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water.

Storage and Stability

Store the lyophilized protein at -20°C to -80°C for long term. After reconstitution, the protein solution is stable at -20°C for 3 months, at 2-8 °C for up to 1 week. This product is stable at ≤ -70°C for up to 6 months from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Data presented is the available current product information and provided as-is. These products have not been tested or verified in any additional applications, sample types, including any clinical use. Experimental conditions must be empirically derived by the user. Our Antibody Guarantee only covers tested applications stated herein and conditions presented in our product information and is not extended to publications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service