Skip to Content
Merck
All Photos(2)

Key Documents

SAB5700178

Sigma-Aldrich

Anti-Asymmetric DiMethyl-Histone H3-R26 antibody produced in rabbit

Synonym(s):

H3/j, H3FJ, HIST1H3J

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€370.00

€370.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€370.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€370.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

15

species reactivity

rat, human, mouse

concentration

0.47 mg/ml

technique(s)

immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200
immunoprecipitation (IP): 1:50-1:200
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

dimethylation (Arg26)

Gene Information

human ... H3C12(8356)

General description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H3 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the small histone gene cluster on chromosome 6p22-p21.3.

Immunogen

A synthetic methylated peptide corresponding to residues surrounding Arg26 of human histone H3

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service