Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300353

Sigma-Aldrich

Monoclonal Anti-CASP8 antibody produced in mouse

clone 1H11, ascites fluid

Synonym(s):

ALPS2B, CAP4, FLICE, MACH, MCH5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€491.00

€491.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€491.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€491.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1H11, monoclonal

mol wt

26 kDa

species reactivity

human, mouse, rat, monkey

technique(s)

direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
immunohistochemistry: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CASP8(841)

Immunogen

Purified recombinant fragment of human CASP8 expressed in E.coli.
Mouse monoclonal antibody raised against CASP8

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changyou Li et al.
Molecular oncology, 8(7), 1220-1230 (2014-05-13)
Little is known regarding molecular markers in head and neck squamous cell carcinoma (HNSCC) that predict responsiveness to different therapeutic regimens or predict HNSCC progression. Mutations in procaspase-8 occur in 9% of HNSCC primary tumors, but the functional consequences of
Hanghui Wang et al.
International journal of oncology, 45(1), 157-164 (2014-04-17)
The specific and efficient delivery of small interfering RNA (siRNA) into cancer cells in vivo remains a major obstacle. In this study, we investigated whether ultrasound-targeted microbubble destruction (UTMD) combined with dual targeting of HSP72 and HSC70 in prostate cancer cell
Wei Zuo et al.
Neuropharmacology, 86, 103-115 (2014-07-16)
Mitochondria dysfunction is implicated in diverse conditions, including metabolic and neurodegenerative disorders. Mitochondrial dynamics has attracted increasing attention as to its relationship with mitochondria autophagy, also known as mitophagy, which is critical for degradation of dysfunctional mitochondria maintaining mitochondrial homeostasis.
Sireesha V Garimella et al.
Breast cancer research : BCR, 16(2), R41-R41 (2014-04-22)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) binds to its receptors, TRAIL-receptor 1 (TRAIL-R1) and TRAIL-receptor 2 (TRAIL-R2), leading to apoptosis by activation of caspase-8 and the downstream executioner caspases, caspase-3 and caspase-7 (caspase-3/7). Triple-negative breast cancer (TNBC) cell lines with
Ulrike Heinicke et al.
Cancer letters, 351(1), 50-58 (2014-05-13)
Rhabdomyosarcoma (RMS) is the most common soft-tissue sarcoma in childhood with a dismal prognosis, highlighting the need for novel treatment strategies. Here, we identify a novel synthetic lethal interaction between the histone deacetylase inhibitor (HDACI) SAHA and anticancer drugs in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service