Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300042

Sigma-Aldrich

Monoclonal Anti-PAR1 antibody produced in mouse

clone 6A7H10, ascites fluid

Synonym(s):

D15S227E, PAR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6A7H10, monoclonal

species reactivity

human

technique(s)

direct ELISA: 1:10,000
western blot: 1:500-1:2,000

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAR1(145624)

Immunogen

Purified recombinant fragment of PAR1 expressed in E.coli.
Mouse monoclonal antibody raised against PAR1

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong-Hua Fu et al.
Molecular medicine reports, 16(5), 7375-7381 (2017-09-26)
The aim of the present study was to determine the effects of early anticoagulation treatment on severe burns complicated by inhalation injury in a rabbit model. Under anesthetization, an electrical burns instrument (100˚C) was used to scald the backs of
Daniela Brünnert et al.
Journal of reproductive immunology, 132, 35-41 (2019-03-25)
Extravillous trophoblast (EVT) migration and invasion is the crucial step for normal placental development. IL-11 is a cytokine regulating cell migration and invasion in cells and is a critical factor for successful implantation of an embryo. Higher expression of thrombin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service