SAB5200154
Monoclonal Anti-Alginate - Atto 594 antibody produced in mouse
clone 3G4-1F5, purified immunoglobulin
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
Atto 594 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
3G4-1F5, monoclonal
form
buffered aqueous solution
species reactivity
wide range
concentration
1 mg/mL
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Specificity
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service