Skip to Content
Merck
All Photos(1)

Key Documents

SAB5200108

Sigma-Aldrich

Anti-SLC14A2 antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Slc14a2, Anti-UT-A1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€417.00

€417.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€417.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 127 kDa
antigen predicted mol wt 97 kDa

species reactivity

rat, mouse

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 14 member 2 (SLC14A2) is composed of 929 amino acids and is expressed in the collecting duct of the inner medulla in rat kidneys. The gene ID of the protein is 54302.

Specificity

Detects ~97 and 127 kDa.

Immunogen

AA911-929 of the rat sequence

Biochem/physiol Actions

Solute carrier family 14 member 2 (SLC14A2) plays a crucial role in increasing the permeability of urea in the collecting ducts of the inner medulla in kidneys of rats. It also helps to maintain the concentration of urine.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoxiao Wang et al.
Scientific reports, 9(1), 18102-18102 (2019-12-04)
Acute liver failure is divided into hyperacute, acute and subacute liver failure. Ascites is a common complication of subacute liver failure. Although animal models of acute liver failure have been established, the study of the pathogenesis of subacute liver failure
Abinash C Mistry et al.
American journal of physiology. Cell physiology, 298(6), C1431-C1437 (2010-05-12)
Of the three major protein variants produced by the UT-A gene (UT-A1, UT-A2, and UT-A3) UT-A1 is the largest. It contains UT-A3 as its NH(2)-terminal half and UT-A2 as its COOH-terminal half. When being part of UT-A1, UT-A3 and UT-A2
Hua Su et al.
American journal of physiology. Renal physiology, 303(2), F201-F208 (2012-04-27)
Urea transporters UT-A1 and UT-A3 are both expressed in the kidney inner medulla. However, the function of UT-A3 remains unclear. Here, we found that UT-A3, which comprises only the NH(2)-terminal half of UT-A1, has a higher urea transport activity than

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service