Skip to Content
Merck
All Photos(1)

Key Documents

SAB5200007

Sigma-Aldrich

Monoclonal Anti-YDJ1 antibody produced in mouse

clone 1G10.H8, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-CPR3, Anti-DNAJ, Anti-DNAJA2, Anti-HIRIP4, Anti-HIRa interatcting protein4, Anti-Hsp40, YDJ1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€417.00

€417.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€417.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G10.H8, monoclonal

form

solution

mol wt

antigen predicted mol wt 40 kDa

species reactivity

yeast

concentration

1 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

yeast ... YDJ1(855661)

Specificity

Detects ~40 kDa.

Immunogen

Full length protein yeast Hsp40 (YDJ1)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in 50% glycerol, and 0.09% sodium azide
aqueous glycerol solution

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michela Ciccarelli et al.
Molecular biology of the cell, 34(10), ar101-ar101 (2023-07-19)
Heat Shock Factor 1 (Hsf1) in yeast drives the basal transcription of key proteostasis factors and its activity is induced as part of the core heat shock response. Exploring Hsf1 specific functions has been challenging due to the essential nature
Anna Maria Eisele-Bürger et al.
Cell reports, 42(11), 113372-113372 (2023-11-08)
Metacaspases are ancestral homologs of caspases that can either promote cell death or confer cytoprotection. Furthermore, yeast (Saccharomyces cerevisiae) metacaspase Mca1 possesses dual biochemical activity: proteolytic activity causing cell death and cytoprotective, co-chaperone-like activity retarding replicative aging. The molecular mechanism
Deepika Gaur et al.
PLoS genetics, 18(11), e1010442-e1010442 (2022-11-10)
Hsp90 constitutes one of the major chaperone machinery in the cell. The Hsp70 assists Hsp90 in its client maturation though the underlying basis of the Hsp70 role remains to be explored. In the present study, using S. cerevisiae strain expressing
Deepika Gaur et al.
Genetics, 215(3), 683-698 (2020-04-18)
Heat-shock protein (Hsp) 90 assists in the folding of diverse sets of client proteins including kinases and growth hormone receptors. Hsp70 plays a major role in many Hsp90 functions by interacting and modulating conformation of its substrates before being transferred

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service