Skip to Content
Merck
All Photos(4)

Key Documents

SAB4702105

Monoclonal Anti-CD200 PE produced in mouse

new

Synonym(s):

MOX, MRC, OX-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

phycoerythrin (R-PE) conjugate

Quality Level

antibody product type

primary antibodies

clone

OX-104, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1κ

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD200(4345)

General description

CD200 (also known as OX2 or MRC) is a type-1 membrane glycoprotein, which contains two extracellular immunoglobulin domains, transmembrane domain and cytoplasmic domain. It is expressed by neuronal cells, B and T cell subsets, follicular dendritic cells, keratinocytes, and ovarian cells. The interaction between CD200 and its receptor CD200R results in macrophage activation (IL-6 production), inhibition of mast cell degranulation along with reduced TNF-alpha and IL-13 secretion and overall attenuation of the activation status of lymphocytes. It seems CD200 is also involved in maternal tolerance and its decreased expression in hair follicle correlates with follicular miniaturization.

Specificity

The mouse monoclonal antibody OX-104 recognizes an extracellular epitope of CD200, a type-1 glycoprotein of the immunoglobulin superfamily, which is expressed in neurons, B and T cell subsets, keratinocytes, follicular dendritic cells, and ovarian cells.

Immunogen

recombinant human CD200

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with R-phycoerythrin (PE) under optimum conditions. Unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service