Skip to Content
Merck
All Photos(1)

Documents

SAB4702100

Monoclonal Anti-Myeloperoxidase PE produced in mouse

new

Synonym(s):

MPO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

phycoerythrin (R-PE) conjugate

Quality Level

antibody product type

primary antibodies

clone

MPO421-8B2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MPO(4353)

General description

Myeloperoxidase (MPO) is a heme enzyme that is localized in azurophilic (primary) granules of myeloid cells and its synthesis occurs at an early stage of differentiation. The mature myeloperoxidase is a tetramer composed of two light (12 kDa) and two heavy (60 kDa) chains. This enzyme uses hydrogen peroxide to oxidize numerous substrates, including serotonin, melatonin or chloride, to produce reactive free radicals that contribute to immune reactions of myeloid cells against pathogens. Myeloperoxidase functions not only in host defense by mediating efficient microbial killing but also can contribute to progressive tissue damage in chronic inflammatory states such as atherosclerosis or acute pancreatitis.

Specificity

The mouse monoclonal antibody MPO421-8B2 recognizes human myeloperoxidase, a heme protein present in intracellular granules of myeloblasts, neutrophils and monocytes. It is a marker of acute myelogenous leukemias and acute lymphoblastic leukemias.

Immunogen

Human myeloperoxidase

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with R-phycoerythrin (PE) under optimum conditions. Unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service