Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700673

Sigma-Aldrich

Monoclonal Anti-Blood Group ABH antibody produced in mouse

clone HE-10, culture supernatant, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€190.00

€190.00


Please contact Customer Service for Availability


Select a Size

Change View
1 ML
€190.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€190.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

HE-10, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: suitable

isotype

IgM

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

The antibody HE-10 agglutinates erythrocytes of group A but does not agglutinate erythrocytes of group B and 0. Study with specific oligosaccharides showed that the antibody HE-10 reacts with A and H antigens with chain types 3 and 4 and it does not react with A disaccharide, A trisaccharide, A type 1, A type 2, ALeb.
The antibody HE-10 does not react with normal tissue sections of donors with blood group B and 0 but it reacts specifically with malignant tissues.

Immunogen

Mixture of erythrocytes of group A1 and glycoprotein fraction isolated from saliva of secretors with blood group A.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in high glucose Dulbecco′s modified Eagle´s Medium (DMEM) with stable glutamine, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service