Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700555

Sigma-Aldrich

Monoclonal Anti-Cd3e-FITC antibody produced in hamster

clone 145-2C11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€277.00

€277.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€277.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

€277.00


Please contact Customer Service for Availability

biological source

hamster

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

145-2C11, monoclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

0.5 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Cd3e(12501)

Related Categories

General description

The hamster monoclonal antibody 145-2C11 reacts with mouse CD3 (epsilon subunit). This antibody is commonly used as a phenotypic marker for mouse T cells.

Immunogen

Mouse BM10-37 cytotoxic T lymphocytes

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 2 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Sempé et al.
Diabetologia, 37(4), 337-343 (1994-04-01)
The NOD mouse, which shows many features of human IDDM, is extensively used to evaluate the role of T lymphocytes in the pathogenesis of autoimmune diabetes. The development of diabetes in this model appears to be controlled by a finely
Takashi Hiromatsu et al.
The Journal of infectious diseases, 187(9), 1442-1451 (2003-04-30)
Interleukin (IL)-15, a potent inhibitor of tumor necrosis factor (TNF)-alpha-mediated apoptosis, causes multiple organ failure during endotoxic shock. We investigated the potential role of IL-15 in protection against Escherichia coli-induced shock by using IL-15 transgenic (Tg) mice. These mice were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service