Skip to Content
Merck
All Photos(1)

Documents

SAB4700552

Sigma-Aldrich

Monoclonal Anti-Cd3e antibody produced in hamster

clone 145-2C11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

hamster

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

145-2C11, monoclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

mouse ... Cd3e(12501)

General description

The hamster monoclonal antibody 145-2C11 reacts with mouse CD3 (epsilon subunit). This antibody is commonly used as a phenotypic marker for mouse T cells.

Immunogen

Mouse BM10-37 cytotoxic T lymphocytes

Application

The reagent is designed for Flow Cytometry analysis using 1-2 μg reagent / 1e6 cells in a suspension.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susanne M A Lens et al.
Molecular and cellular biology, 22(15), 5419-5433 (2002-07-09)
The caspase 8 inhibitor c-FLIP(L) can act in vitro as a molecular switch between cell death and growth signals transmitted by the death receptor Fas (CD95). To elucidate its function in vivo, transgenic mice were generated that overexpress c-FLIP(L) in
Lisa A Pitcher et al.
European journal of immunology, 35(12), 3643-3654 (2005-11-01)
T cell receptor (TCR) signal transduction is mediated by the immunoreceptor tyrosine-based activation motifs (ITAM). The ten ITAM in the TCR complex are distributed in two distinct signaling modules termed TCR zetazeta and CD3 gammaepsilon/deltaepsilon. To delineate the specific role
Qizhi Tang et al.
Journal of immunology (Baltimore, Md. : 1950), 168(9), 4480-4487 (2002-04-24)
FcR nonbinding anti-CD3 epsilon mAbs elicit partial TCR signaling that leads to T cell unresponsiveness and tolerance in vivo. In this study, the membrane-proximal events that promote T cell inactivation by FcR nonbinding anti-CD3 mAbs were examined. In the context
M Hanief Sofi et al.
Molecular immunology, 46(7), 1443-1450 (2009-02-03)
T helper (Th) cell differentiation is a key event to mount an appropriate immune response. Th1 and Th2 cells produce their signature cytokines IFN-gamma and IL-4, respectively. However, as we have reported, CD4 T cells selected by MHC class II-expressing
Marcel Wuthrich et al.
The Journal of experimental medicine, 197(11), 1405-1416 (2003-06-05)
Systemic fungal infections with primary and opportunistic pathogens have become increasingly common and represent a growing health menace in patients with AIDS and other immune deficiencies. T lymphocyte immunity, in particular the CD4+ Th 1 cells, is considered the main

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service