Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700525

Sigma-Aldrich

Monoclonal Anti-VIM-PE antibody produced in mouse

clone VI-RE/1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Vimentin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€406.00

€406.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€406.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

phycoerythrin (R-PE) conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

VI-RE/1, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

0.1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... VIM(7431)

General description

The antibody VI-RE/1 reacts with human vimentin, a 57 kDa intermediate filament protein expressed on a wide variety of mesenchymal and mesodermal cell types.

Immunogen

Bacterially expressed full-length human vimentin

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 10 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service