Skip to Content
Merck
All Photos(6)

Documents

SAB4600066

Sigma-Aldrich

Anti-Mouse IgG (H+L), CF 555 antibody produced in goat

~2 mg/mL, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

CF 555 conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

~2 mg/mL

technique(s)

flow cytometry: 1-10 μg/mL
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect immunofluorescence: 1-10 μg/mL

fluorescence

λex 555 nm; λem 565 nm

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) is a largely expressed serum antibody and is a member of the immunoglobulin family. It consists of gamma (γ) heavy chain in the constant (C) region. Monomeric structure of IgGis composed of two identical heavy chains and two identical light chains with molecular weight of 50kDa and 25kDa respectively. The primary structure of this antibody also contains disulfide bonds implicated in linking the two heavy chains, linking the heavy and light chains and resides inside the chains. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4 with different heavy chains, named γ1,γ2, γ3, and γ4, respectively. IgG is produced and released from CD5+B cells, the primary lymphocytes in the neonatal cell repertoire. Maternal IgG is the only antibody transported across the placenta to the fetus. Maternal IgG passively immunizes the infants.

Specificity

Binds all mouse IgGs

Immunogen

mouse IgG (H+L)

Application

Anti-Mouse IgG (H+L), CF 555 antibody produced in goat has been used in immunohistochemistry and immunofluorescence.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 2 mg/mL bovine serum albumin.

Preparation Note

Protect from light.

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: btinfo@biotium.com.
CF is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mônica Morais-Santos et al.
Endocrinology, 159(2), 723-732 (2017-11-10)
Besides androgens, estrogen signaling plays a key role in normal development and pathologies of the prostate. Irreversible synthesis of estrogens from androgens is catalyzed by aromatase. Interestingly, animals lacking aromatase do not develop cancer or prostatitis, whereas those with overexpression
Basal Cells Show Increased Expression of Aromatase and Estrogen Receptor ? in Prostate Epithelial Lesions of Male Aging Rats
Morais-Santos M
Endocrinology, 159, 723-732 (2018)
Exorhodopsin and melanopsin systems in the pineal complex and brain at early developmental stages of Atlantic halibut (Hippoglossus hippoglossus).
Eilertsen M
The Journal of Comparative Neurology, 522, 4003-4022 (2014)
Yi-Xin Wang et al.
Neural regeneration research, 17(6), 1318-1323 (2021-11-17)
Oscillating field stimulation (OFS) is a potential method for treating spinal cord injury. Although it has been used in spinal cord injury (SCI) therapy in basic and clinical studies, its underlying mechanism and the correlation between its duration and nerve
Mariann Eilertsen et al.
Developmental biology, 444(2), 129-138 (2018-10-22)
In nonmammalian vertebrates, photoreception takes place in the deep brain already early in development, but knowledge is lacking about the functions of these nonvisual photoreceptive systems. Prior to hatching, Atlantic halibut has a transient bilateral cluster of photoreceptive cells in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service