Skip to Content
Merck
All Photos(10)

Key Documents

SAB4504605

Sigma-Aldrich

Anti-phospho-AXL (pTyr698) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ARK, Anti-JTK11, Anti-Tyro7, Anti-UFO

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€459.00

€459.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€459.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€459.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 97 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr698)

Gene Information

human ... AXL(558)

Related Categories

General description

AXL (receptor tyrosine kinase) is also termed as Ark, UFO or Tyro7. This gene is overexpressed in pancreatic cancer tissues. AXL belongs to the receptor tyrosine kinase family. It is obtained as a transforming gene from human leukemia cells. AXL is mapped to human chromosome 19q13.

Immunogen

The antiserum was produced against synthesized peptide derived from human AXL around the phosphorylation site of Tyr691.

Immunogen Range: 657-706

Biochem/physiol Actions

AXL (receptor tyrosine kinase) controls innate immune responses. It participates in cellular transformation and tumor progression. Overexpression of AXL is characterized with higher liability for lymphatic metastases.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Axl mediates ZIKA virus entry in human glial cells and modulates innate immune responses.
Meertens L, et al.
Cell Reports, 18(2), 324-333 (2017)
Victoria H Cruz et al.
JCI insight, 5 (2019-04-03)
Pancreatic ductal adenocarcinoma (PDA) is characterized by an activating mutation in KRAS. Direct inhibition of KRAS through pharmacological means remains a challenge; however, targeting key KRAS effectors has therapeutic potential. We investigated the contribution of TANK-binding kinase 1 (TBK1), a
Yanting Duan et al.
Oncology letters, 17(6), 5784-5792 (2019-06-13)
AXL receptor tyrosine kinase ligand (AXL), a tyrosine kinase receptor that is commonly overexpressed in numerous types of cancer, significantly promotes drug resistance and metastasis in tumor cells. Inhibition of the AXL/growth arrest-specific 6 (Gas6) signaling pathway is emerging as
The Axl receptor tyrosine kinase confers an adverse prognostic influence in pancreatic cancer and represents a new therapeutic target.
Koorstra JBM, et al.
Cancer Biology & Therapy, 8(7), 618-626 (2009)
AXL signaling in primary sensory neurons contributes to chronic compression of dorsal root ganglion-induced neuropathic pain in rats.
Lingli Liang et al.
Molecular pain, 16, 1744806919900814-1744806919900814 (2019-12-31)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service