SAB4503947
Anti-phospho-NFAT4 (pSer165) antibody produced in rabbit
affinity isolated antibody
Select a Size
€390.00
Select a Size
About This Item
€390.00
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 115 kDa
species reactivity
mouse, human, rat
concentration
~1 mg/mL
technique(s)
ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
phosphorylation (pSer165)
Gene Information
human ... NFATC3(4775)
1 of 4
This Item | SAB4504718 | SAB4503946 | SAB4503944 |
---|---|---|---|
conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
mol wt antigen 115 kDa | mol wt antigen 165 kDa | mol wt antigen 95 kDa | mol wt antigen 115 kDa |
UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
Immunogen
Immunogen Range: 131-180
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service