Skip to Content
Merck
All Photos(7)

Key Documents

SAB4503839

Sigma-Aldrich

Anti-phospho-SYK (pTyr525) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€459.00

€459.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€459.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€459.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 72 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr525)

Gene Information

human ... SYK(6850)

General description

Spleen tyrosine kinase (SYK) is a member of the non-receptor tyrosine kinase family. It is usually expressed in epithelial breast tissues and hematopoietic cells. SYK is mapped to human chromosome 9q22.

Immunogen

The antiserum was produced against synthesized peptide derived from human SYK around the phosphorylation site of Tyr525.

Immunogen Range: 491-540

Application

Anti-phospho-SYK (pTyr525) antibody has been used in immunohistochemistry.

Biochem/physiol Actions

Spleen tyrosine kinase (SYK), as a tumor suppressor gene helps to prevent the development and metastasis of breast cancer cells. It participates in B-cell receptor signaling. This protein helps to phosphorylate the α-tubulin subunits of microtubules.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a Novel SYK/c-MYC/MALAT1 Signaling Pathway and Its Potential Therapeutic Value in Ewing Sarcoma
Sun H, et al.
Clinical Cancer Research, 23(15), 4376-4387 (2017)
Conny Tümmler et al.
Cancers, 11(2) (2019-02-13)
Neuroblastoma is a malignancy arising from the developing sympathetic nervous system and the most common and deadly cancer of infancy. New therapies are needed to improve the prognosis for high-risk patients and to reduce toxicity and late effects. Spleen tyrosine
Novel SYK gene variations and changes in binding sites of miRs in breast cancer patients
Faryal R, et al.
Cancer Biomarkers : Section A of Disease Markers, 16(3), 319-326 (2016)
Genomic and expression profiling identifies the B-cell associated tyrosine kinase Syk as a possible therapeutic target in mantle cell lymphoma
Rinaldi A, et al.
British Journal of Haematology, 132(3), 303-316 (2006)
Syk: a new player in the field of breast cancer
Stewart ZA and Pietenpol JA
Breast Cancer Research, 3(1), 5-7 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service