Skip to Content
Merck
All Photos(13)

Documents

SAB4503817

Sigma-Aldrich

Anti-phospho-eEF2 (pThr56) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 95 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EEF2(1938)

General description

Eukaryotic elongation factor 2 (eEF2) is a cytoplasmic protein encoded by the gene mapped to human chromosome 19.

Immunogen

The antiserum was produced against synthesized peptide derived from human eEF2 around the phosphorylation site of Thr56.

Immunogen Range: 31-80

Biochem/physiol Actions

Eukaryotic elongation factor 2 (eEF2) plays a vital role in the elongation of polypeptide chain via catalyzing the translocation of the two tRNAs and the mRNA after peptidyl transfer on the 80S ribosome. eEF2 is highly expressed in various human cancers such as lung, esophageal, pancreatic, breast and prostate cancers, head and neck squamous cell carcinoma (HNSCC), glioblastoma multiforme and non-Hodgkins lymphoma (NHL). eEF2 is used as a potential marker for the diagnosis of non-small cell lung cancer (NSCLC). eEF2 activity is inhibited by the eukaryotic elongation factor-2 kinase (eEF2K), which is essential for the transmission of growth and stress signals to protein synthesis. eEF2 functions as a biochemical sensor in neuronal dendrites that links miniature synaptic events to local translational inactivation. eEF2 is also involved in nuclear deformation and aneuploidy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Jørgensen et al.
Biochemical Society transactions, 34(Pt 1), 1-6 (2005-10-26)
eEF2 (eukaryotic elongation factor 2) occupies an essential role in protein synthesis where it catalyses the translocation of the two tRNAs and the mRNA after peptidyl transfer on the 80 S ribosome. Recent crystal structures of eEF2 and the cryo-electron
Michael A Sutton et al.
Neuron, 55(4), 648-661 (2007-08-19)
Activity-dependent regulation of dendritic protein synthesis is critical for enduring changes in synaptic function, but how the unique features of distinct activity patterns are decoded by the dendritic translation machinery remains poorly understood. Here, we identify eukaryotic elongation factor-2 (eEF2)
Hong-Gang Sun et al.
Asian Pacific journal of cancer prevention : APJCP, 14(11), 6533-6535 (2014-01-01)
The purpose of this study was to evaluate a new type of tumor biomarker, eukaryotic elongation factor 2 (eEF2), in serum for the early diagnosis, confirmative diagnosis as well as assessment of treatment of non-small cell lung cancer (NSCLC). 130
Qi Yao et al.
The Journal of biological chemistry, 289(18), 12666-12678 (2014-03-22)
Protein-tyrosine kinase C-terminal Src kinase (Csk) was originally purified as a kinase for phosphorylating Src and other Src family kinases. The phosphorylation of a C-terminal tyrosine residue of Src family kinases suppresses their kinase activity. Therefore, most physiological studies regarding
Y Kaneda et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(10), 3158-3162 (1984-05-01)
Elongation factor 2 (EF-2), polypeptidyl -tRNA translocase, is an essential factor for protein synthesis in eukaryotic cells and Archebacteria . We isolated diphtheria toxin-resistant human primary embryo cells that contain EF-2 that cannot be ADP-ribosylated by diphtheria toxin and Pseudomonas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service