SAB4502456
Anti-CLTR2 antibody produced in rabbit
affinity isolated antibody
Synonym(s):
CysLTR2, Cysteinyl leukotriene receptor 2, HG57, HPN321, hGPCR21
Select a Size
€391.00
Select a Size
About This Item
€391.00
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 39 kDa
species reactivity
human
concentration
~1 mg/mL
technique(s)
ELISA: 1:5000
western blot: 1:500-1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CYSLTR2(57105)
1 of 4
This Item | SAB4502455 | HPA046528 | SAB2900039 |
---|---|---|---|
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
form buffered aqueous solution | form buffered aqueous solution | form buffered aqueous glycerol solution | form buffered aqueous solution |
clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
General description
Immunogen
Immunogen Range: 281-330
Features and Benefits
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service