SAB4501834
Anti-MARK3 antibody produced in rabbit
affinity isolated antibody
Synonym(s):
C-TAK1, Cdc25C- associated protein kinase 1, MAP/microtubule affinity-regulating kinase 3, Protein kinase STK10, Ser/Thr protein kinase PAR-1
Select a Size
€345.00
Select a Size
About This Item
€345.00
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 86 kDa
species reactivity
mouse, human, rat
concentration
~1 mg/mL
technique(s)
ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MARK3(4140)
Related Categories
General description
The gene MARK3 (microtubule affinity regulating kinase 3) is mapped to human chromosome 14q32.3. It belongs to the Par-1 family of serine/threonine kinases.
Immunogen
Immunogen Range: 1-50
Biochem/physiol Actions
Features and Benefits
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service