Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501252

Sigma-Aldrich

Anti-GPR18 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

G-protein coupled receptor 18, GPCRW, GPR18, N-arachidonyl glycine receptor, NAGly receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€391.00

€391.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€391.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€391.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 38 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR18(2841)

General description

Anti-GPR18 Antibody detects endogenous levels of total GPR18 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human GPR18.

Immunogen Range: 131-180

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lung Inflammation Resolution by RvD1 and RvD2 in a Receptor-Dependent Manner.
Gao, et al.
Pharmaceutics, 15 (2023)
Gang Zuo et al.
Molecular brain, 11(1), 9-9 (2018-02-15)
Cerebral ischemia/reperfusion (I/R) injury is a critical factor leading to a poor prognosis for ischemic stroke patients. ω-3 fatty acid supplements taken as part of a daily diet have been shown to improve the prognosis of patients with ischemic stroke.
Ceren Emre et al.
Acta neuropathologica communications, 9(1), 116-116 (2021-07-01)
Sustained brain chronic inflammation in Alzheimer's disease (AD) includes glial cell activation, an increase in cytokines and chemokines, and lipid mediators (LMs), concomitant with decreased pro-homeostatic mediators. The inflammatory response at the onset of pathology engages activation of pro-resolving, pro-homeostatic LMs followed by
Ceren Emre et al.
Communications biology, 5(1), 245-245 (2022-03-23)
Sustained microglial activation and increased pro-inflammatory signalling cause chronic inflammation and neuronal damage in Alzheimer's disease (AD). Resolution of inflammation follows neutralization of pathogens and is a response to limit damage and promote healing, mediated by pro-resolving lipid mediators (LMs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service