Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501173

Sigma-Aldrich

Anti-Connexin 43 Antibody

rabbit polyclonal

Synonym(s):

CX43, CXA1, CXN-43, Connexin 43, Gap junction 43 kDa heart protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€391.00

€391.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€391.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€391.00


Please contact Customer Service for Availability

Product Name

Anti-Connexin 43 antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 43 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GJA1(2697)

General description

Anti-Connexin 43 Antibody detects endogenous levels of total Connexin 43 protein.
Connexin 43 (Cx43) is also known as GJA1 (gap junction protein α 1) and belongs to a large family of homologous connexin proteins. It has an intracellular N-terminus, four transmembrane domains, two extracellular loops, one cytoplasmic loop and an intracellular C-terminus. It is located on human chromosome 6q22-q23.

Immunogen

The antiserum was produced against synthesized peptide derived from human Connexin 43.

Immunogen Range: 226-275

Application

Anti-Connexin 43 antibody has been used in immunohistochemistry.

Biochem/physiol Actions

Mutations in Connexin 43 (Cx43) gene result in pleiotropic phenotype of oculodentodigital dysplasia. Connexin 43 participates in the multiplication and differentiation of mature T cells. It also participates in the secretion and manufacturing of cytokines. It helps to release lactate from glycolytic pancreatic ductal adenocarcinoma cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spatiotemporal regulation of an Hcn4 enhancer defines a role for Mef2c and HDACs in cardiac electrical patterning
Vedantham V, et al.
Developmental Biology, 373, 149-162 (2013)
Up-regulation of gap junction in peripheral blood T lymphocytes contributes to the inflammatory response in essential hypertension
Ni X, et al.
PLoS ONE, 12 (2017)
Timothy Bage et al.
Archives of dermatological research, 311(8), 647-652 (2019-06-07)
In this study, we characterised the effect that seeding keratinocytes on the papillary and reticular dermis had on the extracellular matrix and tissue integrity ex vivo. Human skin explants from consented patients (n = 6) undergoing routine surgery were cultured at a
Wei Zhang et al.
Behavioral and brain functions : BBF, 19(1), 23-23 (2023-12-19)
As a kind of environmental noise, infrasonic noise has negative effects on various human organs. To date, research has shown that infrasound impairs cognitive function, especially the ability for learning and memory. Previously, we demonstrated that impaired learning and memory
Connexin 43 (GJA1) mutations cause the pleiotropic phenotype of oculodentodigital dysplasia
Paznekas WA, et al.
American Journal of Human Genetics, 72, 408-418 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service