Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500837

Sigma-Aldrich

Anti-PYK2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CADTK, CAK β, Calcium-dependent tyrosine kinase, Cell adhesion kinase β, FADK 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 115 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTK2B(2185)

General description

Anti-PYK2 Antibody detects endogenous levels of total PYK2 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human PYK2.

Immunogen Range: 545-594

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Tong et al.
Physiological research, 68(1), 89-98 (2018-11-16)
The contraction of gastrointestinal (GI) smooth muscles is regulated by both Ca(2+)-dependent and Ca(2+) sensitization mechanisms. Proline-rich tyrosine kinase 2 (Pyk2) is involved in the depolarization-induced contraction of vascular smooth muscle via a Ca(2+) sensitization pathway. However, the role of
Ryan D Mills et al.
The Journal of biological chemistry, 290(14), 8677-8692 (2015-02-26)
Depolarization of the vascular smooth muscle cell membrane evokes a rapid (phasic) contractile response followed by a sustained (tonic) contraction. We showed previously that the sustained contraction involves genistein-sensitive tyrosine phosphorylation upstream of the RhoA/Rho-associated kinase (ROK) pathway leading to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service