Skip to Content
Merck
All Photos(2)

Documents

SAB4500756

Sigma-Aldrich

Anti-APLF antibody produced in rabbit

affinity isolated antibody

Synonym(s):

APFL, C2orf13, Xip1, aprataxin and PNK-like factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 56 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APLF(200558)

General description

Aprataxin and PNKP-like factor (APLF) is also called as PALF and Xip1. It is located in the nucleus. APLF has a forkhead-associated (FHA) domain and two unique poly (ADP-ribose)-binding zinc finger (PBZ) domains in its C-terminal region. This gene is mapped to human chromosome 2p13.3.

Immunogen

The antiserum was produced against synthesized peptide derived from human APLF.

Immunogen Range: 82-131

Biochem/physiol Actions

Aprataxin and PNKP-like factor (APLF) acts as a DNA repair factor. The forkhead-associated (FHA) domain of APLF helps to bind to the phosphorylated FHA domain binding motifs (FBMs) in XRCC1 and XRCC4. Other domains of APLF recruites APLF to sites of DNA damage.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid whole genome sequencing identifies a novel GABRA1 variant associated with West syndrome.
Farnaes L, et al.
Cold Spring Harbor molecular case studies, 3(5), mcs-a001776 (2017)
Characterization of the APLF FHA?XRCC1 phosphopeptide interaction and its structural and functional implications.
Kim K, et al.
Nucleic Acids Research, 45(21), 12374-12387 (2017)
Identification and functional characterization of a Ku-binding motif in aprataxin polynucleotide kinase/phosphatase-like factor (APLF).
Shirodkar P, et al.
The Journal of Biological Chemistry, 288(27), 19604-19613 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service